siRNA Details
Gene Symbol: Eif5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100697.2 | NP_001094167.1 | 3, 4 | 383 | ||
XM_006232231.2 | XP_006232293.1 | 3, 4 | 1414 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eif5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100697.2 | NP_001094167.1 | 3, 4 | 383 | ||
XM_006232231.2 | XP_006232293.1 | 3, 4 | 1414 |