siRNA Details
Gene Symbol: Kcnmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104560.2 | NP_001098030.1 | 4 | 859 | ||
GenBank mRNA | AB297662.1 | - | 4 | 860 |

More Information
Other Species Targeted

Gene Ontology Categories:
