siRNA Details
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107665.1 | NP_001101135.1 | 13 | 1613 | ||
XM_008760924.1 | XP_008759146.1 | 13 | 1697 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107665.1 | NP_001101135.1 | 13 | 1613 | ||
XM_008760924.1 | XP_008759146.1 | 13 | 1697 |