siRNA Details
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107665.1 | NP_001101135.1 | 12 | 1479 | ||
XM_008760924.1 | XP_008759146.1 | 12 | 1563 |

More Information
Other Species Targeted

Gene Ontology Categories:
