siRNA Details
Gene Symbol: LOC102553014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760957.1 | XP_008759179.1 | 4 | 925 | ||
XM_008760958.1 | XP_008759180.1 | 4 | 520 | |||
XM_008760959.1 | XP_008759181.1 | 4 | 1309 | |||
XM_008760960.1 | XP_008759182.1 | 5 | 660 | |||
XM_008760961.1 | XP_008759183.1 | 5 | 645 | |||
XM_008760962.1 | XP_008759184.1 | 4 | 2083 |
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107668.1 | NP_001101138.1 | 4 | 643 | ||
XM_003749267.3 | XP_003749315.1 | 522 | ||||
XM_006232261.2 | XP_006232323.1 | 5 | 641 | |||
XM_006232262.2 | XP_006232324.1 | 4 | 854 | |||
XM_006232263.2 | XP_006232325.1 | 4 | 569 | |||
XM_006232264.2 | XP_006232326.1 | 4 | 1309 | |||
XM_008760925.1 | XP_008759147.1 | 4 | 628 | |||
XM_008760926.1 | XP_008759148.1 | 660 | ||||
XM_008760927.1 | XP_008759149.1 | 4 | 2081 |
