siRNA Details
Gene Symbol: Arse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047885.1 | NP_001041350.1 | 7 | 1045 | ||
XM_006232280.2 | XP_006232342.1 | 8 | 1232 | |||
XM_006232281.2 | XP_006232343.1 | 8 | 1237 | |||
GenBank mRNA | BC168644.1 | - | 6 | 2238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
