siRNA Details
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107669.1 | NP_001101139.1 | 6 | 1735 | ||
XM_006232299.2 | XP_006232361.1 | 7 | 1913 | |||
XM_006232300.1 | XP_006232362.1 | 6 | 1618 | |||
GenBank mRNA | BC166450.1 | - | 7 | 1853 | 8371068 | |
FQ234784.1 | - | 3 | 785 |
