siRNA Details
Gene Symbol: Gpr87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107677.1 | NP_001101147.1 | 1 | 294 | ||
XM_006232401.2 | XP_006232463.1 | 2 | 927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
