siRNA Details
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107680.1 | NP_001101150.1 | 5 | 444 | ||
XM_006232477.2 | XP_006232539.1 | 4 | 407 | |||
XM_006232478.2 | XP_006232540.1 | 345 | ||||
XM_006232479.2 | XP_006232541.1 | 3 | 323 | |||
GenBank mRNA | FQ215154.1 | - | 5 | 447 | ||
FQ215514.1 | - | 5 | 441 | |||
FQ216377.1 | - | 5 | 405 | |||
FQ216411.1 | - | 5 | 473 | |||
FQ223879.1 | - | 5 | 409 | |||
FQ224275.1 | - | 2 | 102 |

More Information
Other Species Targeted

Gene Ontology Categories:
