siRNA Details
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107680.1 | NP_001101150.1 | 2 | 224 | ||
XM_006232477.2 | XP_006232539.1 | 2 | 232 | |||
XM_006232478.2 | XP_006232540.1 | 125 | ||||
XM_006232479.2 | XP_006232541.1 | 2 | 232 | |||
GenBank mRNA | FQ215154.1 | - | 2 | 227 | ||
FQ215514.1 | - | 2 | 221 | |||
FQ216377.1 | - | 2 | 185 | |||
FQ216411.1 | - | 2 | 253 | |||
FQ223879.1 | - | 2 | 189 |
