siRNA Details
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107681.2 | NP_001101151.1 | 4 | 878 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178726, AK147389, AK142789, AK147876, AK045724, and others.
Gene Ontology Categories:
