siRNA Details
Gene Symbol: Nmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107682.2 | NP_001101152.1 | 13, 14 | 1347 | ||
XM_008761090.1 | XP_008759312.1 | 13, 14 | 1345 | |||
GenBank mRNA | BC161998.1 | - | 13, 14 | 1347 | 9035992 | |
FQ215207.1 | - | 11, 12 | 1084 |
