siRNA Details
Gene Symbol: Rapgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232511.2 | XP_006232573.2 | 5, 6 | 2089 | ||
XM_006232512.2 | XP_006232574.1 | 3, 4 | 1155 | |||
XM_006232513.2 | XP_006232575.1 | 3, 4 | 966 | |||
XM_008761092.1 | XP_008759314.1 | 9, 10 | 1459 | |||
XM_008761093.1 | XP_008759315.1 | 8, 9 | 1442 | |||
XM_008761094.1 | XP_008759316.1 | 9, 10 | 1459 | |||
XM_008761095.1 | XP_008759317.1 | 9, 10 | 1133 | |||
XM_008761096.1 | XP_008759318.1 | 7, 8 | 1748 | |||
XM_008761097.1 | XP_008759319.1 | 6, 7 | 1640 | |||
XM_008761098.1 | XP_008759320.1 | 6, 7 | 2932 |
