siRNA Details
Gene Symbol: RGD1560010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107685.1 | NP_001101155.1 | 1, 2 | 264 | ||
GenBank mRNA | BC162053.1 | - | 1, 2 | 228 | 7113318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
