siRNA Details
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033896.1 | NP_001029068.1 | 2 | 401 | ||
XM_006232637.2 | XP_006232699.1 | 2 | 455 | |||
XM_008761149.1 | XP_008759371.1 | 1 | 920 | |||
GenBank mRNA | BC101913.1 | - | 2 | 401 | 7936201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
