siRNA Details
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012078.2 | NP_001012078.2 | 5, 6 | 688 | ||
XM_006232639.2 | XP_006232701.1 | 5, 6 | 568 | |||
XM_006232640.2 | XP_006232702.1 | 5, 6 | 539 | |||
XM_006232641.2 | XP_006232703.1 | 5, 6 | 768 | |||
XM_008761150.1 | XP_008759372.1 | 5, 6 | 1262 | |||
GenBank mRNA | BC081943.1 | - | 5, 6 | 632 | 7121310 |
