siRNA Details
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012079.1 | NP_001012079.1 | 13 | 1703 | ||
XM_006232643.2 | XP_006232705.1 | 18 | 2481 | |||
XM_006232644.2 | XP_006232706.1 | 19 | 2598 | |||
XM_006232646.2 | XP_006232708.1 | 17 | 2058 | |||
XM_006232647.2 | XP_006232709.1 | 16 | 1902 | |||
XM_006232648.2 | XP_006232710.1 | 16 | 1853 | |||
XM_006232651.2 | XP_006232713.1 | 13 | 1991 | |||
XM_006232652.2 | XP_006232714.1 | 13 | 1810 | |||
XM_006232653.2 | XP_006232715.1 | 13 | 1698 | |||
XM_006232654.2 | XP_006232716.1 | 13 | 2507 | |||
XM_008761151.1 | XP_008759373.1 | 19 | 2593 | |||
XM_008761152.1 | XP_008759374.1 | 14 | 2581 | |||
GenBank mRNA | BC090078.1 | - | 13 | 1703 | 7125049 | |
FQ212011.1 | - | 14 | 1676 |
