siRNA Details
Gene Symbol: Ash1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107689.1 | NP_001101159.1 | 3 | 4982 | ||
XM_008761153.1 | XP_008759375.1 | 3 | 4957 | |||
XM_008761154.1 | XP_008759376.1 | 2 | 4825 | |||
XM_008761155.1 | XP_008759377.1 | 3 | 5109 | |||
XM_008761156.1 | XP_008759378.1 | 4 | 5244 | |||
XM_008761157.1 | XP_008759379.1 | 4 | 5130 | |||
XM_008761158.1 | XP_008759380.1 | 3 | 5026 | |||
XM_008761159.1 | XP_008759381.1 | 3 | 4874 | |||
XM_008761160.1 | XP_008759382.1 | 2 | 4825 |
