siRNA Details
Gene Symbol: Efna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107692.1 | NP_001101162.1 | 2 | 457 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007910, AK012195, U90663, AK142604.
Gene Ontology Categories:
