siRNA Details
Gene Symbol: Pmvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008352.1 | NP_001008353.1 | 5 | 865 | ||
GenBank mRNA | BC086349.1 | - | 5 | 865 | 7302798 | |
FQ226470.1 | - | 5 | 995 | |||
FQ233923.1 | - | 5 | 995 | |||
FQ234368.1 | - | 5 | 995 |
