siRNA Details
Gene Symbol: Pogz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107693.1 | NP_001101163.1 | 17 | 2445 | ||
XM_008761295.1 | XP_008759517.1 | 19 | 2817 | |||
XM_008761296.1 | XP_008759518.1 | 19 | 2790 | |||
XM_008761297.1 | XP_008759519.1 | 18 | 2658 | |||
XM_008761298.1 | XP_008759520.1 | 18 | 2631 | |||
XM_008761299.1 | XP_008759521.1 | 18 | 2499 |
