siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271175.1 | NP_001258104.1 | 13 | 2021 | ||
XM_006232913.2 | XP_006232975.1 | 14 | 2411 | |||
XM_006232914.2 | XP_006232976.1 | 13 | 2192 | |||
XM_006232916.2 | XP_006232978.1 | 9 | 1388 | |||
XM_006232917.2 | XP_006232979.1 | 9 | 1891 |
