siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271175.1 | NP_001258104.1 | 2, 3 | 370 | ||
XM_006232913.2 | XP_006232975.1 | 3, 4 | 760 | |||
XM_006232914.2 | XP_006232976.1 | 2, 3 | 541 | |||
GenBank mRNA | DN937937.1 | - | 2, 3 | 398 |
