siRNA Details
Gene Symbol: Tars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014040.1 | NP_001014062.1 | 13, 14 | 1648 | ||
XM_008761308.1 | XP_008759530.1 | 14, 15 | 4167 | |||
GenBank mRNA | BC079154.1 | - | 13, 14 | 1648 | 7128703 | |
FQ232225.1 | - | 5, 6 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
