siRNA Details
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012080.1 | NP_001012080.1 | 4 | 1371 | ||
GenBank mRNA | BC089203.1 | - | 4 | 1371 | 7191112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
