siRNA Details
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012080.1 | NP_001012080.1 | 3, 4 | 864 | ||
GenBank mRNA | BC089203.1 | - | 3, 4 | 864 | 7191112 |

More Information
Other Species Targeted

Gene Ontology Categories:
