siRNA Details
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009691.3 | NP_001009691.3 | 16 | 2624 | ||
NM_001195832.1 | NP_001182761.1 | 16 | 2624 | |||
XM_006232679.2 | XP_006232741.1 | 16 | 2794 | |||
XM_006232680.2 | XP_006232742.1 | 16 | 2791 | |||
XM_006232681.2 | XP_006232743.1 | 15 | 2745 | |||
XM_006232682.2 | XP_006232744.1 | 15 | 2742 | |||
XM_006232684.2 | XP_006232746.1 | 16 | 2794 | |||
XM_006232685.2 | XP_006232747.1 | 15 | 2741 | |||
XM_008761176.1 | XP_008759398.1 | 15 | 2714 | |||
XM_008761177.1 | XP_008759399.1 | 16 | 1689 | |||
GenBank mRNA | AY673997.1 | - | 16 | 2016 | ||
AY673998.1 | - | 16 | 2096 | |||
AY674000.1 | - | 16 | 2016 |
