siRNA Details
Gene Symbol: Fam46c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014041.1 | NP_001014063.1 | 2 | 821 | ||
XM_006233038.2 | XP_006233100.1 | 2 | 639 | |||
XM_006233039.2 | XP_006233101.1 | 2 | 743 | |||
XM_008761361.1 | XP_008759583.1 | 2 | 611 | |||
GenBank mRNA | BC083570.1 | - | 2 | 821 | 7111947 |

More Information
Other Species Targeted

