siRNA Details
Gene Symbol: Cd101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271217.1 | NP_001258146.1 | 6 | 2052 | ||
XM_006233049.2 | XP_006233111.1 | 5 | 1637 | |||
XM_008761362.1 | XP_008759584.1 | 6 | 2822 |

More Information
Other Species Targeted

Gene Ontology Categories:
