siRNA Details
Gene Symbol: Fam212b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107713.1 | NP_001101183.1 | 2 | 806 | ||
XM_006233088.2 | XP_006233150.1 | 3 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
