siRNA Details
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233199.2 | XP_006233261.1 | 3, 4 | 310 | ||
XM_006233200.2 | XP_006233262.1 | 3, 4 | 310 | |||
XM_006233204.2 | XP_006233266.1 | 3, 4 | 1673 | |||
XM_008761446.1 | XP_008759668.1 | 4, 5 | 433 | |||
GenBank mRNA | BC098000.1 | - | 3, 4 | 189 |
