siRNA Details
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107717.1 | NP_001101187.1 | 1, 2 | 375 | ||
XM_006233159.2 | XP_006233221.1 | 2, 3 | 383 | |||
XM_006233160.2 | XP_006233222.1 | 2, 3 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
