siRNA Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127544.1 | NP_001121016.1 | 6 | 799 | ||
XM_008761404.1 | XP_008759626.1 | 5 | 827 | |||
GenBank mRNA | BC162022.1 | - | 6 | 778 | 9098607 | |
FQ211892.1 | - | 5 | 759 | |||
FQ221591.1 | - | 6 | 879 |
