siRNA Details
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191715.1 | NP_001178644.1 | 9 | 1026 | ||
GenBank mRNA | BC091417.1 | - | 5 | 384 | ||
BC107441.1 | - | 5 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
