siRNA Details
Gene Symbol: Extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100704.1 | NP_001094174.1 | 5 | 966 | ||
XM_006233215.2 | XP_006233277.1 | 5 | 1511 | |||
XM_006233216.2 | XP_006233278.1 | 5 | 1252 | |||
XM_006233217.2 | XP_006233279.1 | 4 | 1043 | |||
GenBank mRNA | BC091373.1 | - | 1 | 110 | ||
BC162014.1 | - | 5 | 966 | 9091843 |
