siRNA Details
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107718.2 | NP_001101188.2 | 11 | 1715 | ||
NM_001134856.1 | NP_001128328.1 | 11 | 1641 | |||
XM_008761460.1 | XP_008759682.1 | 12 | 1165 | |||
GenBank mRNA | BC161876.1 | - | 11 | 1715 | 7115002 |
