siRNA Details
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135871.1 | NP_001129343.1 | 8 | 719 | ||
XM_006233327.2 | XP_006233389.1 | 8 | 2316 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146141, AK143475, AK003660, AK090384, BC011417.
Gene Ontology Categories:
