siRNA Details
Gene Symbol: Ppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135871.1 | NP_001129343.1 | 9 | 833 | ||
XM_006233327.2 | XP_006233389.1 | 9 | 2430 |

More Information
Other Species Targeted

Gene Ontology Categories:
