siRNA Details
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031655.1 | NP_001026825.1 | 7 | 931 | ||
GenBank mRNA | BC099094.1 | - | 7 | 931 | 7458970 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031655.1 | NP_001026825.1 | 7 | 931 | ||
GenBank mRNA | BC099094.1 | - | 7 | 931 | 7458970 |