siRNA Details
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031655.1 | NP_001026825.1 | 11, 12 | 1492 | ||
GenBank mRNA | BC099094.1 | - | 11, 12 | 1492 | 7458970 | |
BC111710.1 | - | 3, 4 | 452 | 7383173 |
