siRNA Details
Gene Symbol: Alpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761482.1 | XP_008759704.1 | 10 | 1522 | ||
XM_008761483.1 | XP_008759705.1 | 9 | 1102 | |||
XM_227715.7 | XP_227715.5 | 9 | 1629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
