siRNA Details
Gene Symbol: Zgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233293.2 | XP_006233355.1 | 10, 11 | 4256 | ||
XM_006233294.2 | XP_006233356.1 | 10, 11 | 3652 | |||
XM_006233296.2 | XP_006233358.1 | 8, 9 | 3942 | |||
XM_006233297.2 | XP_006233359.1 | 5, 6 | 2745 | |||
XM_008761479.1 | XP_008759701.1 | 9, 10 | 4147 | |||
XM_008761480.1 | XP_008759702.1 | 9, 10 | 3569 |
