siRNA Details
Gene Symbol: Zgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233293.2 | XP_006233355.1 | 7 | 3302 | ||
XM_006233294.2 | XP_006233356.1 | 7 | 2698 | |||
XM_006233296.2 | XP_006233358.1 | 6 | 3174 | |||
XM_006233297.2 | XP_006233359.1 | 2 | 1791 | |||
XM_008761479.1 | XP_008759701.1 | 6 | 3193 | |||
XM_008761480.1 | XP_008759702.1 | 6 | 3173 |
