siRNA Details
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107728.1 | NP_001101198.1 | 4, 5 | 491 | ||
XM_006233395.2 | XP_006233457.1 | 2, 3 | 2551 | |||
XM_008761511.1 | XP_008759733.1 | 2, 3 | 2555 | |||
XM_008761512.1 | XP_008759734.1 | 3, 4 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
