siRNA Details
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034926.1 | NP_001030098.1 | 10 | 982 | ||
XM_006233538.2 | XP_006233600.1 | 14 | 1691 | |||
XM_006233539.2 | XP_006233601.1 | 12 | 1353 | |||
XM_008761564.1 | XP_008759786.1 | 13 | 1583 | |||
XM_008761565.1 | XP_008759787.1 | 12 | 3295 | |||
XM_008761566.1 | XP_008759788.1 | 13 | 2473 | |||
GenBank mRNA | BC088311.1 | - | 2 | 159 | ||
BC105888.1 | - | 10 | 982 | 7308716 | ||
FQ212592.1 | - | 4 | 355 |
