siRNA Details
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034926.1 | NP_001030098.1 | 5 | 428 | ||
XM_006233538.2 | XP_006233600.1 | 9 | 1137 | |||
XM_006233539.2 | XP_006233601.1 | 7 | 799 | |||
XM_008761564.1 | XP_008759786.1 | 9 | 1137 | |||
XM_008761565.1 | XP_008759787.1 | 7 | 2741 | |||
XM_008761566.1 | XP_008759788.1 | 8 | 1919 | |||
GenBank mRNA | BC105888.1 | - | 5 | 428 | 7308716 |
