siRNA Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134549.2 | NP_001128021.2 | 9 | 2416 | ||
XM_006233489.2 | XP_006233551.1 | 11 | 2700 | |||
XM_006233490.2 | XP_006233552.1 | 12 | 3290 | |||
XM_006233491.2 | XP_006233553.1 | 11 | 2697 | |||
XM_008761551.1 | XP_008759773.1 | 12 | 2729 | |||
GenBank mRNA | FM103952.1 | - | 7 | 605 | ||
FN806367.1 | - | 6 | 579 |

More Information
Other Species Targeted

Gene Ontology Categories:
