siRNA Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134549.2 | NP_001128021.2 | 7 | 2199 | ||
XM_006233489.2 | XP_006233551.1 | 9 | 2483 | |||
XM_006233490.2 | XP_006233552.1 | 10 | 3073 | |||
XM_006233491.2 | XP_006233553.1 | 9 | 2480 | |||
XM_008761551.1 | XP_008759773.1 | 10 | 2512 | |||
GenBank mRNA | FM103952.1 | - | 5 | 388 | ||
FN806367.1 | - | 4 | 362 | |||
FQ227845.1 | - | 1 | 590 |

More Information
Other Species Targeted

Gene Ontology Categories:
