siRNA Details
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191094.1 | NP_001178023.1 | 23 | 2578 | ||
XM_006233492.2 | XP_006233554.1 | 24 | 2779 | |||
XM_006233493.2 | XP_006233555.1 | 24 | 2755 | |||
GenBank mRNA | BC092624.1 | - | 3 | 506 |
