siRNA Details
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191094.1 | NP_001178023.1 | 13 | 1581 | ||
XM_006233492.2 | XP_006233554.1 | 14 | 1770 | |||
XM_006233493.2 | XP_006233555.1 | 14 | 1746 |

More Information
Other Species Targeted

Gene Ontology Categories:
